Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZEB2
Entrez Gene ID
539873
Gene Name
zinc finger E-box binding homeobox 2
Gene Aliases
ZFHX1B
Chromosome Location
Chr.2: 52174189 - 52264481 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076192.2 NP_001069660.2 1 - 2 520 70
GenBank mRNA BC120151.1 - 1 - 2 178 70

View Full Product Details