Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXB9
Entrez Gene ID
783873
Gene Name
homeobox B9
Gene Aliases
-
Chromosome Location
Chr.19: 37866983 - 37870934 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191186.1 NP_001178115.1 1 - 2 576 64

View Full Product Details