Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VEGFA
Entrez Gene ID
281572
Gene Name
vascular endothelial growth factor A
Gene Aliases
VEGF, VEGF-A, VPF, eVEGF120, eVEGF164
Chromosome Location
Chr.23: 17281915 - 17297193 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001316955.1 NP_001303884.1 6 - 7 1602 83
NM_001316956.1 NP_001303885.1 7 - 8 1674 83
NM_174216.2 NP_776641.1 6 - 7 1602 83
GenBank mRNA GU362062.1 - 6 - 7 555 83
M31836.1 - 5 - 6 477 83
M32976.1 - 6 - 7 1100 83
MK424008.1 - 1 - 2 145 83

View Full Product Details