Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTCHD1
Entrez Gene ID
536443
Gene Name
patched domain containing 1
Gene Aliases
-
Chromosome Location
Chr.X: 119819982 - 119873093 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192881.1 NP_001179810.1 2 - 3 1016 114

View Full Product Details