Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MOGAT1
Entrez Gene ID
407225
Gene Name
monoacylglycerol O-acyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.2: 110843551 - 110878526 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001153.2 NP_001001153.1 2 - 3 273 80
GenBank mRNA AJ519785.2 - 2 - 3 273 80

View Full Product Details