Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGA5
Entrez Gene ID
414350
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
pga
Chromosome Location
Chr.29: 39760936 - 39771371 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001600.2 NP_001001600.2 7 - 8 955 60
GenBank mRNA AY330769.1 - 6 - 7 876 60
BC149644.1 - 6 - 7 905 60

View Full Product Details