Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK11B
Entrez Gene ID
493708
Gene Name
cyclin dependent kinase 11B
Gene Aliases
CDC2L1, CDC2L2, CDK11
Chromosome Location
Chr.16: 50983533 - 51003446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007811.2 NP_001007812.2 12 - 13 1393 97
GenBank mRNA AY788896.1 - 3 - 4 313 97

View Full Product Details