Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TKDP2
Entrez Gene ID
503553
Gene Name
trophoblast Kunitz domain protein 2
Gene Aliases
-
Chromosome Location
Chr.13: 74145316 - 74155090 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012683.2 NP_001012701.1 5 - 6 525 96
GenBank mRNA AF241777.1 - 5 - 6 526 96

View Full Product Details