Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FDFT1
Entrez Gene ID
281767
Gene Name
farnesyl-diphosphate farnesyltransferase 1
Gene Aliases
-
Chromosome Location
Chr.8: 7587994 - 7617580 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013004.1 NP_001013022.1 3 - 4 486 99
GenBank mRNA BC109959.1 - 3 - 4 567 99

View Full Product Details