Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKBIL1
Entrez Gene ID
514369
Gene Name
NFKB inhibitor like 1
Gene Aliases
-
Chromosome Location
Chr.23: 27732883 - 27743646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015597.1 NP_001015597.1 1 - 2 138 136
GenBank mRNA BT020645.1 - 1 - 2 138 136
BT021709.1 - 1 - 2 122 136

View Full Product Details