Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP18
Entrez Gene ID
515202
Gene Name
ubiquitin specific peptidase 18
Gene Aliases
bubp43
Chromosome Location
Chr.5: 76001952 - 76018301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017940.1 NP_001017940.1 8 - 9 924 78
GenBank mRNA AJ865078.1 - 8 - 9 924 78
BC149487.1 - 9 - 10 1085 78

View Full Product Details