Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC51
Entrez Gene ID
515066
Gene Name
leucine-rich repeat-containing protein 51
Gene Aliases
-
Chromosome Location
Chr.15: 51714865 - 51729483 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024545.1 NP_001019716.1 5 - 6 650 82
NM_001282038.1 NP_001268967.1 5 - 6 611 82
NM_001282039.1 NP_001268968.1 4 - 5 550 82
GenBank mRNA BC109501.1 - 4 - 5 551 82
BT020631.1 - 5 - 6 650 82
EE348175.1 - 5 - 6 611 82

View Full Product Details