Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBM3
Entrez Gene ID
509771
Gene Name
RNA binding motif protein 3
Gene Aliases
-
Chromosome Location
Chr.X: 86690789 - 86694645 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303463.1 NP_001290392.1 4 - 5 421 116
NM_001303480.1 NP_001290409.1 4 - 5 421 116
GenBank mRNA BC103473.1 - 4 - 5 911 116
DT860071.1 - 4 - 5 407 116
EE239224.1 - 4 - 5 381 116

View Full Product Details