Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM3
Entrez Gene ID
509771
Gene Name
RNA binding motif protein 3
Gene Aliases
-
Chromosome Location
Chr.X: 86690789 - 86694645 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC103473.1 - 5 - 6 1023 134

View Full Product Details