Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDH1
Entrez Gene ID
534090
Gene Name
3-hydroxybutyrate dehydrogenase 1
Gene Aliases
-
Chromosome Location
Chr.1: 71964466 - 72000614 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034600.2 NP_001029772.1 3 - 4 378 57
GenBank mRNA BC103051.1 - 3 - 4 379 57

View Full Product Details