Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA7
Entrez Gene ID
616809
Gene Name
golgin A7
Gene Aliases
-
Chromosome Location
Chr.27: 36419388 - 36436498 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035484.1 NP_001030561.1 2 - 3 295 133
GenBank mRNA BC120011.1 - 3 - 4 344 133
BT020714.1 - 2 - 3 295 133

View Full Product Details