Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THRSP
Entrez Gene ID
515940
Gene Name
thyroid hormone responsive
Gene Aliases
-
Chromosome Location
Chr.29: 17989040 - 17994946 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040533.1 NP_001035623.1 1 - 2 505 61
GenBank mRNA AY656814.1 - 1 - 2 505 61
BC114894.1 - 1 - 2 494 61

View Full Product Details