Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPC1
Entrez Gene ID
767977
Gene Name
mitochondrial pyruvate carrier 1
Gene Aliases
BRP44L
Chromosome Location
Chr.9: 101357079 - 101370413 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077042.2 NP_001070510.1 3 - 4 305 90
GenBank mRNA BC102377.1 - 3 - 4 306 90

View Full Product Details