Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COCH
Entrez Gene ID
504316
Gene Name
cochlin
Gene Aliases
-
Chromosome Location
Chr.21: 41526381 - 41540310 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077842.1 NP_001071310.1 7 - 8 576 108
GenBank mRNA BC123841.1 - 7 - 8 579 108
BT020705.1 - 7 - 8 576 108

View Full Product Details