Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPE
Entrez Gene ID
280753
Gene Name
carboxypeptidase E
Gene Aliases
CPH
Chromosome Location
Chr.17: 573369 - 721446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173903.4 NP_776328.2 6 - 7 1361 109
GenBank mRNA BC142181.1 - 6 - 7 1227 109
CB437028.1 - 2 - 3 320 109
EW682509.1 - 3 - 4 276 109
MK423995.1 - 2 - 3 280 109
X04411.1 - - 992 109

View Full Product Details