Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZP3
Entrez Gene ID
280964
Gene Name
zona pellucida glycoprotein 3
Gene Aliases
Zp-3, Zp-3B
Chromosome Location
Chr.25: 34446154 - 34453895 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173974.3 NP_776399.2 1 - 2 337 59
GenBank mRNA BC146035.1 - 1 - 2 339 59
BT021613.1 - 1 - 2 261 59
U05775.1 - 1 - 2 454 59

View Full Product Details