Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLIN2
Entrez Gene ID
280981
Gene Name
perilipin 2
Gene Aliases
ADFP
Chromosome Location
Chr.8: 25117110 - 25131174 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173980.2 NP_776405.1 2 - 3 91 118
GenBank mRNA AF541976.1 - 2 - 3 91 118
AJ011680.1 - 2 - 3 91 118
BC102211.1 - 2 - 3 113 118
BT029909.1 - 2 - 3 101 118

View Full Product Details