Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASR
Entrez Gene ID
281038
Gene Name
calcium sensing receptor
Gene Aliases
BoPCaR1
Chromosome Location
Chr.1: 66655318 - 66744722 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174002.3 NP_776427.1 4 - 5 1888 60
GenBank mRNA S67307.1 - 4 - 5 1892 60

View Full Product Details