Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSF3
Entrez Gene ID
281096
Gene Name
colony stimulating factor 3
Gene Aliases
-
Chromosome Location
Chr.19: 40329796 - 40332910 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174028.1 NP_776453.1 2 - 3 217 103
GenBank mRNA AF092533.1 - 2 - 3 217 103

View Full Product Details