Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2K
Entrez Gene ID
281225
Gene Name
ubiquitin conjugating enzyme E2 K
Gene Aliases
E2-25K, HIP2, HYPG, LIG
Chromosome Location
Chr.6: 58774538 - 58854226 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174080.2 NP_776505.1 6 - 7 550 118
GenBank mRNA BC142324.1 - 6 - 7 713 118
S51016.1 - 6 - 7 550 118

View Full Product Details