Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IBSP
Entrez Gene ID
281233
Gene Name
integrin binding sialoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 36879556 - 36893855 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174084.3 NP_776509.1 3 - 4 138 142
GenBank mRNA S73144.1 - 3 - 4 142 142

View Full Product Details