Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFNG
Entrez Gene ID
281237
Gene Name
interferon gamma
Gene Aliases
-
Chromosome Location
Chr.5: 45624513 - 45629336 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174086.1 NP_776511.1 3 - 4 456 73
GenBank mRNA EF675629.1 - 3 - 4 363 73
EU276066.1 - 3 - 4 386 73
EU277737.1 - 3 - 4 363 73
FJ263670.1 - 3 - 4 363 73
M29867.1 - 3 - 4 456 73

View Full Product Details