Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC12
Entrez Gene ID
281259
Gene Name
DnaJ heat shock protein family (Hsp40) member C12
Gene Aliases
JDP1
Chromosome Location
Chr.28: 24248418 - 24296785 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174096.2 NP_776521.1 4 - 5 648 106
GenBank mRNA AB042112.1 - 4 - 5 648 106
BC142111.1 - 4 - 5 106
BC149977.1 - 5 - 6 106

View Full Product Details