Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDYN
Entrez Gene ID
281385
Gene Name
prodynorphin
Gene Aliases
-
Chromosome Location
Chr.13: 53130235 - 53148819 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174139.3 NP_776564.2 2 - 3 307 145
GenBank mRNA BC151343.1 - 2 - 3 292 145
U58500.1 - 2 - 3 305 145

View Full Product Details