Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXN2
Entrez Gene ID
281557
Gene Name
thioredoxin 2
Gene Aliases
-
Chromosome Location
Chr.5: 74883639 - 74893619 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174208.2 NP_776633.1 3 - 4 454 93
GenBank mRNA BC112876.1 - 3 - 4 484 93
D87741.1 - 3 - 4 454 93

View Full Product Details