Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CABP1
Entrez Gene ID
281653
Gene Name
calcium binding protein 1
Gene Aliases
-
Chromosome Location
Chr.17: 62859837 - 62919832 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174254.2 NP_776679.1 5 - 6 561 70
GenBank mRNA AF169150.1 - 4 - 5 384 70
AF169151.1 - 5 - 6 561 70

View Full Product Details