Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIC5
Entrez Gene ID
281696
Gene Name
chloride intracellular channel 5
Gene Aliases
-
Chromosome Location
Chr.23: 19180486 - 19350762 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174276.3 NP_776701.1 5 - 6 1298 75
GenBank mRNA AF109199.1 - 5 - 6 1192 75
L16547.1 - 5 - 6 1299 75

View Full Product Details