Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLTB
Entrez Gene ID
281698
Gene Name
clathrin light chain B
Gene Aliases
CLTLB
Chromosome Location
Chr.7: 37859120 - 37879373 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174277.2 NP_776702.1 1 - 2 330 126
GenBank mRNA BC148120.1 - 1 - 2 352 126
X04852.1 - - 269 126
X04853.1 - - 330 126

View Full Product Details