Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLTB
Entrez Gene ID
281698
Gene Name
clathrin light chain B
Gene Aliases
CLTLB
Chromosome Location
Chr.7: 37859120 - 37879373 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174277.2 NP_776702.1 4 - 5 602 76
GenBank mRNA X04853.1 - - 602 76

View Full Product Details