Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FABP3
Entrez Gene ID
281758
Gene Name
fatty acid binding protein 3
Gene Aliases
FABP, FABP-3, H-FABP
Chromosome Location
Chr.2: 122287099 - 122294639 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174313.2 NP_776738.1 1 - 2 117 66
GenBank mRNA BC102153.1 - 1 - 2 132 66
FJ756345.1 - 1 - 2 71 66
X12710.1 - - 117 66
X51933.1 - - 108 66

View Full Product Details