Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FABP4
Entrez Gene ID
281759
Gene Name
fatty acid binding protein 4, adipocyte
Gene Aliases
-
Chromosome Location
Chr.14: 44676559 - 44680947 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174314.2 NP_776739.1 2 - 3 304 133
GenBank mRNA X89244.1 - - 304 133

View Full Product Details