Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTIF2
Entrez Gene ID
281923
Gene Name
mitochondrial translational initiation factor 2
Gene Aliases
-
Chromosome Location
Chr.11: 37973499 - 37996167 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174393.2 NP_776818.1 8 - 9 1161 58
GenBank mRNA BC150118.1 - 8 - 9 1156 58
L37835.1 - 8 - 9 1161 58

View Full Product Details