Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OPRM1
Entrez Gene ID
281958
Gene Name
opioid receptor mu 1
Gene Aliases
-
Chromosome Location
Chr.9: 90804291 - 90968682 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174408.2 NP_776833.1 1 - 2 447 87
GenBank mRNA U89677.2 - 1 - 2 447 87

View Full Product Details