Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBA2
Entrez Gene ID
282203
Gene Name
crystallin beta A2
Gene Aliases
-
Chromosome Location
Chr.2: 106933695 - 106936940 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174524.2 NP_776949.1 4 - 5 486 63
GenBank mRNA M60329.1 - 4 - 5 486 63

View Full Product Details