Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRA4
Entrez Gene ID
282238
Gene Name
gamma-aminobutyric acid type A receptor subunit alpha4
Gene Aliases
-
Chromosome Location
Chr.6: 65498266 - 65572998 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174543.2 NP_776968.1 2 - 3 237 107
GenBank mRNA BC123424.1 - 2 - 3 366 107
X61456.1 - 1 - 1 237 107

View Full Product Details