Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRA4
Entrez Gene ID
282238
Gene Name
gamma-aminobutyric acid type A receptor subunit alpha4
Gene Aliases
-
Chromosome Location
Chr.6: 65498266 - 65572998 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174543.2 NP_776968.1 6 - 7 752 88
GenBank mRNA BC123424.1 - 6 - 7 881 88
X61456.1 - - 752 88

View Full Product Details