Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PABPN1
Entrez Gene ID
282298
Gene Name
poly(A) binding protein nuclear 1
Gene Aliases
-
Chromosome Location
Chr.10: 21590460 - 21597349 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174569.2 NP_776994.1 1 - 2 2444 115
GenBank mRNA BC133468.1 - 1 - 2 363 115
BC133551.1 - 2 - 3 496 115
BC133558.1 - 1 - 2 354 115
X89969.1 - - 2444 115

View Full Product Details