Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DYNLT1
Entrez Gene ID
282380
Gene Name
dynein light chain Tctex-type 1
Gene Aliases
TCTEL1
Chromosome Location
Chr.9: 95047826 - 95055238 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174620.2 NP_777045.1 1 - 1 58 77
GenBank mRNA AF067370.1 - 1 - 1 58 77
BC108160.1 - 1 - 2 79 77

View Full Product Details