Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC25A4
Entrez Gene ID
282478
Gene Name
solute carrier family 25 member 4
Gene Aliases
ANT1
Chromosome Location
Chr.27: 15489566 - 15493429 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174658.2 NP_777083.1 3 - 4 742 76
GenBank mRNA BC102994.1 - 3 - 4 839 76
M24102.1 - 3 - 4 744 76

View Full Product Details