Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B14
Entrez Gene ID
282652
Gene Name
hydroxysteroid 17-beta dehydrogenase 14
Gene Aliases
SDR3, retSDR3
Chromosome Location
Chr.18: 55427060 - 55447469 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174701.2 NP_777126.1 1 - 2 158 75
GenBank mRNA AF202997.1 - 1 - 2 158 75
AF238859.1 - 1 - 2 158 75
BC133473.1 - 1 - 2 164 75

View Full Product Details