Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELA2A
Entrez Gene ID
282687
Gene Name
chymotrypsin-like elastase family, member 2A
Gene Aliases
ELA2A, Ela2
Chromosome Location
Chr.16: 52022456 - 52038857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174714.2 NP_777139.1 2 - 3 142 61
GenBank mRNA BC134791.1 - 2 - 3 155 61
X97635.1 - - 142 61

View Full Product Details