Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRYBB2
Entrez Gene ID
287011
Gene Name
crystallin beta B2
Gene Aliases
-
Chromosome Location
Chr.17: 65304388 - 65318606 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174807.1 NP_777232.1 4 - 5 361 131
GenBank mRNA M22466.1 - 4 - 5 361 131

View Full Product Details