Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YWHAZ
Entrez Gene ID
287022
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Gene Aliases
-
Chromosome Location
Chr.14: 63394416 - 63429119 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174814.2 NP_777239.1 3 - 4 474 101
GenBank mRNA BC102382.1 - 3 - 4 508 101
L07955.1 - 3 - 4 474 101

View Full Product Details