Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPING1
Entrez Gene ID
281035
Gene Name
serpin family G member 1
Gene Aliases
C1NH
Chromosome Location
Chr.15: 80849917 - 80863386 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174821.2 NP_777246.1 6 - 7 1150 90
GenBank mRNA U30332.1 - 6 - 7 1150 90

View Full Product Details