Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BSP3
Entrez Gene ID
317695
Gene Name
binder of sperm 3
Gene Aliases
SVS8
Chromosome Location
Chr.18: 51586356 - 51589682 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174840.1 NP_777265.1 4 - 5 454 69
GenBank mRNA AF055981.1 - 4 - 5 454 69

View Full Product Details